Skip to Content
Merck
All Photos(3)

Key Documents

68255

Sigma-Aldrich

4-Methyl-2-oxovaleric acid

≥98.0% (T)

Synonym(s):

α-Ketoisocaproic acid, 2-Oxoisocaproic acid, 4-Methyl-2-oxopentanoic acid, Ketoleucine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCOOH
CAS Number:
Molecular Weight:
130.14
Beilstein:
1701823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥98.0% (T)

refractive index

n20/D 1.431

bp

82-83 °C/11 mmHg (lit.)

mp

8-10 °C

density

1.055 g/mL at 20 °C (lit.)

functional group

carboxylic acid
ketone

storage temp.

2-8°C

SMILES string

CC(C)CC(=O)C(O)=O

InChI

1S/C6H10O3/c1-4(2)3-5(7)6(8)9/h4H,3H2,1-2H3,(H,8,9)

InChI key

BKAJNAXTPSGJCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-2-oxovaleric acid, also known as α-ketoisocaproic acid (KIC), is a metabolite of leucine. [1]It is commonly used as a building block in the synthesis of leucine and its derivatives, and it plays a role in amination reactions and asymmetric synthesis.[2] Additionally, KIC serves as an intermediate in the formation of Strecker aldehydes.[3]

Application

  • Role in Human Brain Ketometabolism: 4-Methyl-2-oxovaleric acid has been implicated in the temporal patterns of ketometabolism in cerebral microdialysis fluids of patients with traumatic brain injury, underscoring its significance in biochemical pathways within the brain (Eiden et al., 2019).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yulan Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(34), 12676-12681 (2004-08-18)
Schistosomiasis, a chronic and debilitating parasitic disease, affects approximately 200 million people in the developing world and imposes a substantial public health and economic impact. Accurately diagnosing at the individual level, monitoring disease progression, and assessing the impact of pharmacological
The First Highly Enantioselective Homogeneously Catalyzed Asymmetric Reductive Amination:? Synthesis of ?-N-Benzylamino Acids
Kadyrov et al.
The Journal of Organic Chemistry, 68, 4067-4070 (2003)
Intermediate role of ?-keto acids in the formation of Strecker aldehydes
Hidalgo et al.
Basic Food Chemistry, 141, 1140-1146 (2013)
Lisa S Chow et al.
American journal of physiology. Endocrinology and metabolism, 291(4), E729-E736 (2006-05-18)
Despite being an anabolic hormone in skeletal muscle, insulin's anticatabolic mechanism in humans remains controversial, with contradictory reports showing either stimulation of protein synthesis (PS) or inhibition of protein breakdown (PB) by insulin. Earlier measurements of muscle PS and PB
W E Mitch et al.
The Journal of clinical investigation, 67(2), 553-562 (1981-02-01)
We measured the effects of seven consecutive daily infusions of alpha-ketoisocaproate (the alpha-keto analogue of leucine) or leucine itself on urinary urea and total nitrogen excretion during fasting. Two study protocols were undertaken. In protocol I, subjects underwent three separate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service