Skip to Content
Merck
All Photos(1)

Key Documents

681806

Sigma-Aldrich

Ammonium lauryl sulfate solution

~30% in H2O

Synonym(s):

Ammonium dodecyl sulfate, Dodecyl sulfate ammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3NH4
CAS Number:
Molecular Weight:
283.43
Beilstein:
5188379
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

concentration

~30% in H2O

Quality Level

refractive index

n20/D 1.374

density

1.02 g/mL at 25 °C

SMILES string

N.CCCCCCCCCCCCOS(O)(=O)=O

InChI

1S/C12H26O4S.H3N/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);1H3

InChI key

BTBJBAZGXNKLQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium lauryl sulfate is an anionic surfactant which can be used in the preparation of porous building ceramics by gelcasting and formulation of cosmetic products. It can also be used as a corrosion inhibitor for carbon steel in acidic solution.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of surfactants on the microstructure and some intrinsic properties of porous building ceramics fabricated by gelcasting.
Fuji M, et al.
Ceramics International, 32(7), 797-802 (2006)
Physico-chemical in vitro methods for determination of the skin compatibility of surfactants.
Zeidler U
Journal of Society of Cosmetic Chemists of Japan, 20(1), 17-26 (1986)
The viscosity and zeta potential of bentonite dispersions in presence of anionic surfactants.
Yalc?n T, et al.
Materials Letters, 57(2), 420-424 (2002)
Sunita,?ADS, ALS, AHDS and ADDBS Surfactants as Corrosion Inhibitors for Carbon Steel in acidic Solution?.
Harish K
Research Journal of Chemical Sciences, 2(7), 1-6 (2012)
Dusty B Brown et al.
Journal of bacteriology, 193(18), 4766-4778 (2011-07-19)
Rhizobium leguminosarum is a Gram-negative bacterium that forms nitrogen-fixing symbioses with compatible leguminous plants via intracellular invasion and establishes a persistent infection within host membrane-derived subcellular compartments. Notably, an unusual very-long-chain fatty acid (VLCFA) is found in the lipid A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service