Skip to Content
Merck
All Photos(1)

Documents

681059

Sigma-Aldrich

2-Methyl-6-nitrobenzoic anhydride

97%

Synonym(s):

MNBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12N2O7
CAS Number:
Molecular Weight:
344.28
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

173-177 °C

SMILES string

Cc1cccc(c1C(=O)OC(=O)c2c(C)cccc2[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C16H12N2O7/c1-9-5-3-7-11(17(21)22)13(9)15(19)25-16(20)14-10(2)6-4-8-12(14)18(23)24/h3-8H,1-2H3

InChI key

YEKPNMQQSPHKBP-UHFFFAOYSA-N

General description

2-Methyl-6-nitrobenzoic anhydride is a reagent employed as a coupling promoter in the synthesis of amides, lactones, esters, and peptides.

Application

2-Methyl-6-nitrobenzoic anhydride can be used:
  • As a versatile lactonization reagent applicable in the preparation of varieties of macrolide natural products and lactones.
  • As a reaction promoter in the synthesis of carboxamide derivatives by using corresponding amines and carboxylic acids.
  • In the total synthesis of GRP78 inhibitor prunustatin A, antifungal compound (3R,16E,20E,23R)-(−)-eushearilide and an antiobestic drug tetrahydrolipstatin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Methyl-6-nitrobenzoic anhydride
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2009)
Takayuki Tonoi et al.
ACS omega, 6(5), 3571-3577 (2021-02-16)
A depsipeptidic analogue of FE399 was efficiently synthesized mainly through macrolactamization using 2-methyl-6-nitrobenzoic anhydride (MNBA), and a detailed investigation of the desired 16-membered macrolactam core of FE399 was performed. It was determined that the combination of MNBA and a catalytic
Ryohei Hirano et al.
Journal of mass spectrometry : JMS, 53(8), 665-674 (2018-05-17)
CYP3A phenotyping provides a means for personalized drug therapy. We focused our attention on the plasma 6β-hydroxycortisol (6β-OHF) to cortisol ratio as an index for CYP3A phenotyping. In the present study, we developed a sensitive and reliable method for the
Dahye Kim et al.
Molecules (Basel, Switzerland), 26(4) (2021-02-12)
Steroid 5-α reductase (5AR) is responsible for the reduction of steroids to 5-α reduced metabolites, such as the reduction of testosterone to 5-α dihydrotestosterone (DHT). A new adverse outcome pathway (AOP) for 5AR inhibition to reduce female reproduction in fish
The first total synthesis of (-) and (+)-2-hydroxy-24-oxooctacosanolide using an effective lactonization
Shiina I, et al.
Organic Letters, 8(21), 4955-4958 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service