Skip to Content
Merck
All Photos(2)

Key Documents

680850

Sigma-Aldrich

Di-(4-chlorobenzyl)azodicarboxylate

97%

Synonym(s):

Bis(4-chlorobenzyl)azodicarboxylate, DCAD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12Cl2N2O4
CAS Number:
Molecular Weight:
367.18
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

108-112 °C

functional group

azo
chloro

SMILES string

O=C(/N=N\C(OCC1=CC=C(Cl)C=C1)=O)OCC2=CC=C(Cl)C=C2

InChI

1S/C16H12Cl2N2O4/c17-13-5-1-11(2-6-13)9-23-15(21)19-20-16(22)24-10-12-3-7-14(18)8-4-12/h1-8H,9-10H2/b20-19-

InChI key

UIFGGABIJBWRMG-VXPUYCOJSA-N

General description

Learn More at the Professor and Product Portal of Professor Bruce Lipshutz.

Application

Di-(4-chlorobenzyl)azodicarboxylate (DCAD) is a novel, stable, solid alternative to DEAD and DIAD for a variety of Mitsunobu couplings giving a readily separable hydrazine byproduct that can be recycled.
Reactant for preparation of:
  • Amino thioesters via guanidine-catalyzed biomimetic enantioselective decarboxylative Mannich and amination reactions of malonic acid half thioesters
  • Hydroacylation reaction of aldehydes in Ionic liquid (IL) medium
  • DCAD (di-p-chlorobenzyl azodicarboxylate) for Mitsunobu coupling reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

T Amirabadi Farahani et al.
Journal of dairy science, 100(8), 6199-6217 (2017-05-30)
This experiment was conducted to compare conventional (CON; 21 d) and shortened (SH; 10 d) close-up period, and evaluate the effect of shortened close-up period combined with feeding different metabolizable protein (MP) levels on dry matter (DM) intake, metabolic status
Kyohei Ishida et al.
Animal science journal = Nihon chikusan Gakkaiho, 89(1), 132-139 (2017-09-16)
Lowering dietary cation-anion differences (DCAD) can enhance responsiveness to Ca-homeostatic hormones and increase Ca availability, which might have potential to activate a Ca-dependent protease, calpain, and to enhance postmortem myofibrillar proteolysis. In this study, we investigated the effects of DCAD
Vivian F Su et al.
BMC developmental biology, 7, 43-43 (2007-05-09)
The Hedgehog (Hh) family of secreted growth factors are morphogens that act in development to direct growth and patterning. Mutations in human Hh and other Hh pathway components have been linked to human diseases. Analysis of Hh distribution during development
M R Wilkens et al.
Journal of animal physiology and animal nutrition, 100(1), 156-166 (2015-02-04)
The concept of feeding anionic salts in late gestation is widely used to prevent milk fever in dairy cows. While the effects of these diets on renal Ca excretion and tissue responsiveness towards parathyroid hormone have clearly been demonstrated, data
Z Wang et al.
Journal of applied microbiology, 130(3), 722-735 (2020-08-07)
The effect of increasing dietary cation-anion difference (DCAD) on rumen fermentation and ruminal microbial community in dairy cows under heat stress (HS) conditions were evaluated. This study was performed as a two-period cross-over design during the summer season, with eight

Related Content

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Designer surfactants enable diverse transformations like Suzuki-Miyaura, Olefin Metathesis, and 1,4-Addition to Enones in water by Prof. Bruce Lipshutz.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service