Skip to Content
Merck
All Photos(3)

Key Documents

679704

Sigma-Aldrich

3′-Hydroxy-4′-methoxyacetophenone

97%

Synonym(s):

1-(3-hydroxy-4-methoxyphenyl)ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

88-92 °C

functional group

ketone

SMILES string

COc1ccc(cc1O)C(C)=O

InChI

1S/C9H10O3/c1-6(10)7-3-4-9(12-2)8(11)5-7/h3-5,11H,1-2H3

InChI key

YLTGFGDODHXMFB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Z Sun et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 18(6), 362-363 (1993-06-01)
3-Hydroxy-4-methoxy-acetophenone (HMA) isolated from Cynanchum paniculatum was found to have analgesic effect and inhibitory action on the gastro-intestinal motility. The effect and action were equivalent to that of paeonol, HMA displayed a very low degree of antibacterial activity against Escherichia
Lee Fielding et al.
Magnetic resonance in chemistry : MRC, 49(7), 405-412 (2011-05-07)
The association constants for the interactions of 2-hydroxy-4-methoxyacetophenone, 2-hydroxy-5-methoxyacetophenone, 2-hydroxy-6-methoxyacetophenone, 3-hydroxy-4-methoxyacetophenone and 4-hydroxy-3-methoxyacetophenone with β-cyclodextrin in water were measured by (1)H NMR and by isothermal titration calorimetry. Very good agreement was obtained between the different methods. The errors associated with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service