Skip to Content
Merck
All Photos(1)

Documents

671231

Sigma-Aldrich

Trifluoroethylene

≥97.5% (GC)

Synonym(s):

Trifluoroethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FCH=CF2
CAS Number:
Molecular Weight:
82.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.5% (GC)

SMILES string

F\C=C(/F)F

InChI

1S/C2HF3/c3-1-2(4)5/h1H

InChI key

MIZLGWKEZAPEFJ-UHFFFAOYSA-N

Application

Trifluoroethylene (TrFE) can be used as a monomer unit for the preparation of fluoropolymer (poly(trifluoroethylene)), which finds application in microelectronics, protective/anti-adhesive coatings, chemical industry and Li-ion batteries. TrFE is also used to synthesize copolymer composites with vinylidene difluoride for various applications.
It can also be used to prepare:
  • Pentafluoroethane by reacting with xenon difluoride.
  • Lithium (trifluorovinyl)trimethoxy borate by treating with trimethyl borate and n-BuLi.
  • 2,2,3-Trifluorooxirane.

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1B - Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen O Leung et al.
The Journal of chemical physics, 128(6), 064315-064315 (2008-02-20)
Guided by ab initio calculations, Fourier transform microwave rotational spectra in the 6-22 GHz region are obtained for the complex formed between 1,1,2-trifluoroethylene and acetylene, including the normal isotopomer, three of four singly substituted (13)C species obtained in natural abundance
Shao-Ling Yang et al.
Canadian Urological Association journal = Journal de l'Association des urologues du Canada, 7(11-12), E681-E686 (2013-11-28)
Our objective is to assess the effects of low-frequency ultrasound combined with microbubbles on benign prostate hyperplasia (BPH). Sixteen Beagle dogs with BPH were randomly assigned into 4 groups (n = 4): control group (without treatment), G1 group (injection with
Reusability of photocatalytic TiO2 and ZnO nanoparticles immobilized in poly (vinylidene difluoride)-co-trifluoroethylene
Teixeira S, et al.
Applied Surface Science, 384(7), 497-504 (2016)
D H Turnbull et al.
Ultrasound in medicine & biology, 21(1), 79-88 (1995-01-01)
There is a growing interest in high resolution, subsurface imaging of cutaneous tissues using higher frequency ultrasound, and several commercial systems have been developed recently which operate at 20 MHz. Some of the possible applications of higher frequency skin imaging
M T Baker et al.
Toxicology and applied pharmacology, 114(1), 25-30 (1992-05-11)
The effect of isoflurane on trifluoroethene (TFE) defluorination and cytochrome P450 inactivation was examined in rats to determine the influence of this anesthetic on in vivo fluoroethene metabolism. Exposure of rats to TFE (0.5%, v/v) or a mixture of TFE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service