Skip to Content
Merck
All Photos(2)

Key Documents

669431

Sigma-Aldrich

Lithium fluoride

Precipitated, 99.995%

Synonym(s):

Fluorolithium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiF
CAS Number:
Molecular Weight:
25.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

99.995%

form

powder

impurities

≤55.0 ppm Trace Metal Analysis

mp

845 °C (lit.)

density

2.64 g/mL at 25 °C (lit.)

SMILES string

[Li+].[F-]

InChI

1S/FH.Li/h1H;/q;+1/p-1

InChI key

PQXKHYXIUOZZFA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium fluoride is an inorganic halide with good ionic conductivity, high refractive index, and high transmittance in the UV region. Owing to its unique properties, it is widely used in the field of optical and electronic devices, X-ray imaging, and dental materials.[1] [2]

Application

Lithium fluoride(LiF) can be used:
  • To fabricate an insulating layer in AC-driven organic light-emitting diodes(OLEDs). LiF provides excellent capacitance and high stability to the insulating layers in AC devices.[1]
  • As a precursor to synthesize Eu2+/Ce3+ doped barium lithium fluoride phosphors by solvothermal method.[2]
  • To fabricate LiF thin film coating on the lithium anodes to enhance their stability and coulombic efficiency.[3]
  • As an additive to enrich and enhance the performance of solidelectrolyte interface(SEI) in Li metal batteries.[4]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alternating current driven organic light emitting diodes using lithium fluoride insulating layers
Shang-Yi Liu, et al.
Scientific Reports, 4, 7559-7559 (2014)
The intrinsic behavior of lithium fluoride in solid electrolyte interphases on lithium
Mingfu He, et al.
Proceedings of the National Academy of Sciences, 117, 73-79 (2020)
Solvothermal fabrication and luminescent properties of Eu2+/Ce3+ doped barium lithium fluoride
Guoxian ZHU, et al.
Journal of Rare Earths, 30, 985-989 (2012)
Novel ALD Chemistry Enabled Low-Temperature Synthesis of Lithium Fluoride Coatings for Durable Lithium Anodes
Lin Chen, et al.
ACS Applied Materials & Interfaces, 10, 26972-26981 (2018)
X W Meng et al.
The journal of physical chemistry. B, 115(16), 4768-4773 (2011-04-06)
On the basis of molecular dynamics simulations, we investigate water permeation across a single-walled carbon nanotube (SWCNT) under the influence of four symmetrical half-rings, each having six LiF dipolar molecules. The flux remains almost fixed as the separation, R, between

Articles

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

Professor Gogotsi and Dr. Shuck introduce MXenes: a promising family of two-dimensional materials with a unique combination of high conductivity, hydrophilicity, and extensive tunability.

See All

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service