Skip to Content
Merck
All Photos(1)

Documents

668257

Sigma-Aldrich

3,4-Dimethoxythiophene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2S
CAS Number:
Molecular Weight:
144.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

refractive index

n20/D 1.5409

bp

100-102 °C/10-11 mmHg

density

1.209 g/mL at 25 °C

storage temp.

−20°C

SMILES string

COc1cscc1OC

InChI

1S/C6H8O2S/c1-7-5-3-9-4-6(5)8-2/h3-4H,1-2H3

InChI key

ZUDCKLVMBAXBIF-UHFFFAOYSA-N

General description

3,4-Dimethoxythiophene (DMOT) is a monomer and a precursor which can be synthesized by ring closure reaction of 2,3-dimethoxy-1,3-butadiene and sulfur dichloride in hexane medium. It is an oligothiphene that is majorly used in the development of electroactive materials for organic electronics based applications.

Application

Building block in the synthesis of an N2S2-N4 porphyrin dyad used to study photoinduced energy transfer.
DMOT can be trans-esterified to form 3,4-ethylenendioxythiophene (EDOT). It can further be polymerized to produce PEDOT which can be used as a conductive polymer in π-conjugated systems. It can be polymerized to form poly(dimethoxythiphenes) which can potentially be used in the fabrication energy storage devices on electrochemical doping.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.1 °F - closed cup

Flash Point(C)

106.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Revisiting the electropolymerization of 3, 4-dimethoxythiophene in organic and micellar media.
Fall M, et al.
Synthetic Metals, 123(3), 365-372 (2001)
Thieno [3, 4-b]-1, 4-oxathiane: An Unsymmetrical Sulfur Analogue of 3, 4-Ethylenedioxythiophene (EDOT) as a Building Block for Linear pi-Conjugated Systems.
Blanchard P, et al.
Organic Letters, 4(4), 607-609 (2002)
Biomimetic Synthesis of Water Soluble Conductive Polypyrrole and Poly (3, 4 ethylenedioxythiophene).
Bruno FF, et al.
MRS Online Proceedings Library, 736(4), 607-609 (2002)
Sokkalingam Punidha et al.
The Journal of organic chemistry, 73(1), 323-326 (2007-12-12)
Click chemistry has been successfully applied in the synthesis of the first example of a triazole-bridged porphyrin dyad containing N(2)S(2) porphyrin and N(4) or ZnN(4) porphyrin subunits, and fluorescence study indicated a possibility of singlet-singlet energy transfer from the N(4)
In situ conductance studies of p-and n-doping of poly (3, 4-dialkoxythiophenes).
Skompska M, et al.
Journal of Electroanalytical Chemistry, 577(1), 9-17 (2005)

Articles

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Organic electronics promise renewable energy solutions surpassing silicon-based tech.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service