Skip to Content
Merck
All Photos(1)

Key Documents

667943

Sigma-Aldrich

Hafnium(IV) n-butoxide

Synonym(s):

n-Butoxy hafnium, Tetrabutoxyhafnium(IV)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H36HfO4
CAS Number:
Molecular Weight:
470.94
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid

Quality Level

reaction suitability

core: hafnium
reagent type: catalyst

density

1.2376 g/mL

SMILES string

CCCCO[Hf](OCCCC)(OCCCC)OCCCC

InChI

1S/4C4H9O.Hf/c4*1-2-3-4-5;/h4*2-4H2,1H3;/q4*-1;+4

InChI key

LTBRWBUKPWVGFA-UHFFFAOYSA-N

General description

Hafnium(IV) n-butoxide is an organometal oxide widely used in the preparation of Hafnium nanomaterials and as a catalyst in polymerization reactions.[1] [2]

Application

Hafnium(IV) n-butoxide can be used:
  • As a sol-gel precursor to synthesize hafnium oxide(HfO2) ultra-thin films with enhanced dielectric properties.[1]
  • As a transesterification catalyst for the synthesis of poly(butylene succinate) (PBS).[2]
  • As a precursor to fabricate BaHfO3(Barium hafnate) perovskitenanocrystals for various electronic applications.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hybrid approach to obtain high-quality BaMO3 perovskite nanocrystals
Natalia Chamorro, et al.
Royal Society of Chemistry Advances, 10, 28872-28878 (2020)
Sol?Gel Fabrication of Dielectric HfO2 Nano-Films; Formation of Uniform, Void-Free Layers and Their Superior Electrical Properties
Yoshitaka Aoki, et al.
Chemistry of Materials, 17, 450-458 (2005)
Synthesis and properties of poly (butylene succinate): Efficiency of different transesterification catalysts
Nicolas Jacquel, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 49 , 5301-5312 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service