Skip to Content
Merck
All Photos(1)

Key Documents

663913

Sigma-Aldrich

N,N′-Dioctyl-3,4,9,10-perylenedicarboximide

98%

Synonym(s):

PTCDI-C8

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H42N2O4
CAS Number:
Molecular Weight:
614.77
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

>300 °C

λmax

526 nm

fluorescence

λem ≤533 nm in chloroform

semiconductor properties

N-type (mobility=1.7 cm2/V·s)

SMILES string

CCCCCCCCN1C(=O)c2ccc3c4ccc5C(=O)N(CCCCCCCC)C(=O)c6ccc(c7ccc(C1=O)c2c37)c4c56

InChI

1S/C40H42N2O4/c1-3-5-7-9-11-13-23-41-37(43)29-19-15-25-27-17-21-31-36-32(40(46)42(39(31)45)24-14-12-10-8-6-4-2)22-18-28(34(27)36)26-16-20-30(38(41)44)35(29)33(25)26/h15-22H,3-14,23-24H2,1-2H3

InChI key

YFGMQDNQVFJKTR-UHFFFAOYSA-N

Application

PTCDI-C8 can be used as an organic semiconductor to fabricate a wide range of opto-electronic based devices such as light emitting diodes, photovoltaic cells, and field effect transistors.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiali Zhang et al.
Nanoscale horizons, 5(7), 1096-1105 (2020-05-20)
Ultrathin organic semiconductor (OSC) crystalline films hold the promise of achieving high-performance, flexible, and transparent organic electronic devices. However, fast and high-throughput solution deposition of uniform pinhole-free ultrathin OSC crystalline films over a large area remains a challenge. Here, we
Three-dimensional molecular packing of thin organic films of PTCDI-C8 determined by surface X-ray diffraction.
Krauss TN, et al.
Langmuir, 24(22), 12742-12744 (2008)
Organic photovoltaic devices based on pentacene/N, N′ -dioctyl-3, 4, 9, 10-perylenedicarboximide heterojunctions.
Karak S, et al.
Organic Electronics, 10(5), 1006-1010 (2009)
N Hiroshiba et al.
Physical chemistry chemical physics : PCCP, 13(13), 6280-6285 (2011-03-02)
Photo-induced carrier processes at the heteromolecular interface of N,N'-dioctyl-3,4,9,10-perylenedicarboximide (PTCDI-C(8)) and quaterrylene (QT) on a molecular scale were examined by optical and photoelectron spectroscopy. The energy level alignments of the molecules were determined by X-ray photoelectron spectroscopy and the optical
Synthesis of PTCDI-C8 one dimensional nanostructures for photovoltaic applications.
IOP Conference Series: Materials Science and Engineering, 73(1), 012052-012052 (2015)

Articles

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

Flexible electronic circuits, displays, and sensors based on organic active materials will enable future generations of electronics products that may eventually enter the mainstream electronics market.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service