Skip to Content
Merck
All Photos(3)

Key Documents

663212

Sigma-Aldrich

Isopropenylboronic acid pinacol ester

contains phenothiazine as stabilizer, 95%

Synonym(s):

2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(isopropenyl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-(1-methylethenyl)-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17BO2
CAS Number:
Molecular Weight:
168.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

contains

phenothiazine as stabilizer

refractive index

n20/D 1.4320

bp

47-49 °C/9 mbar

density

0.894 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=C)B1OC(C)(C)C(C)(C)O1

InChI

1S/C9H17BO2/c1-7(2)10-11-8(3,4)9(5,6)12-10/h1H2,2-6H3

InChI key

SVSUYEJKNSMKKW-UHFFFAOYSA-N

Application

Reagent used for
  • Palladium-catalyzed Suzuki-Miyaura cross-coupling processes[1]
  • Inverse-electron-demand Diels-Alder reaction[2]
  • Simmons-Smith Cyclopropanation Reaction[3]
  • Polyene cyclization[4]
  • Stereoselective aldol reactions[5]
  • Grubbs cross-metathesis reaction[5]
  • Intramolecular Suzuki-Miyaura reaction[5]
  • Stereoselective cross-metathesis[6]
  • Dipolar cycloaddition[7]
  • Iodosulfonylation[8]
  • Asymmetric conjugate addition and intramolecular hydroacylation[9]

Reagent used in preparation of various therapeutic kinase and enzymatic inhibitors[10]
Reagent used for
  • Palladium-catalyzed Suzuki-Miyaura cross-coupling reactions
  • Inverse-electron-demand Diels-Alder reaction
  • Simmons-Smith Cyclopropanation Reaction
  • Polyene cyclization
  • Stereoselective aldol reactions
  • Grubbs cross-metathesis reaction
  • Intramolecular Suzuki-Miyaura reaction
  • Stereoselective cross-metathesis
  • Dipolar cycloaddition
  • Iodosulfonylation
  • Asymmetric conjugate addition and intramolecular hydroacylation

Reagent used in preparation of various therapeutic kinase and enzymatic inhibitors

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F

Flash Point(C)

42 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Jun Zhao et al.
Organic letters, 10(11), 2143-2145 (2008-04-29)
The first asymmetric total synthesis of antiochic acid using bioinspired polyene cyclization strategy is described. Both good yield and good asymmetric induction were obtained.
Catalytic asymmetric intramolecular hydroacylation with rhodium/phosphoramidite-alkene ligand complexes.
Thomas J Hoffman et al.
Angewandte Chemie (International ed. in English), 50(45), 10670-10674 (2011-09-20)
Simmons-Smith cyclopropanation reaction
Charette, A. B.; Beauchemin, A.
Org. React., 58 (2001)
Facile Access to 3,5-Dihalogenated Pyrazoles by Sydnone Cycloaddition and their Versatile Functionalization by Pd-Catalyzed Cross-Coupling Processes
Delaunay, T.; et al.
European Journal of Organic Chemistry, 20-21, 3837-3848 (2011)
First Total Synthesis of Rhodexin A
Jung, M. E.; Yoo, D.
Organic Letters, 13, 3766-3766 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service