Skip to Content
Merck
All Photos(2)

Key Documents

659452

Sigma-Aldrich

1-Adamantanethiol

95%

Synonym(s):

1-AD, 1-AdSH, 1-AdmSH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16S
CAS Number:
Molecular Weight:
168.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

solid

mp

99-106 °C

SMILES string

SC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C10H16S/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9,11H,1-6H2/t7-,8+,9-,10-

InChI key

ADJJLNODXLXTIH-CHIWXEEVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Adamantanethiol (AT) is a bulky caged hydrocarbon based thiol that has no free rotational configurations in its backbone. It forms a self-assembled monolayer (SAM) by attaching order close packed structured to the surface atoms.

Application

AT can be used to functionalize folic acid based polyethylene glycol-maleimide which can be used in the development of gold nanorod (AuNR) based mixtures for spatiotemporal delivery of anticancer therapeutics. It can also be used with 1-octadecanethiol which forms SAM on palladium/aluminum oxide (Pd/Al2O3) which finds potential application as a controlled catalyst.
Reagent used to prepare 1-adamantylthio sialosides for glycosylation with mono- and oligosaccharides on treatment with NIS in triflic acid.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal study and structural characterization of self-assembled monolayers generated from diadamantane disulfide on Au (1 1 1)
Azzam W, et al.
Applied Surface Science, 257(8), 3739-3747 (2011)
A dual wavelength-activatable gold nanorod complex for synergistic cancer treatment
Pacardo DB, et al.
Nanoscale, 7(28), 12096-12103 (2015)
David Crich et al.
The Journal of organic chemistry, 72(20), 7794-7797 (2007-09-11)
Novel 1-adamantanylthio sialosides were synthesized and coupled to acceptors under NIS/TfOH promotion conditions. These donors showed higher reactivity than the phenylthio sialosides and could be activated by NIS/TfOH in nitrile solvents at -78 degrees C to afford improved alpha-sialylations. With
Control of Pd catalyst selectivity with mixed thiolate monolayers
Lien C and Medlin JW
J. Catal., 339(28), 38-46 (2016)
Arrelaine A Dameron et al.
Journal of the American Chemical Society, 127(24), 8697-8704 (2005-06-16)
We have designed monolayers with weak intermolecular interactions for use as placeholders in intelligent self- and directed-assembly. We have shown that these 1-adamantanethiolate monolayers are labile with respect to displacement by exposing them to dilute solutions of alkanethiols. These self-assembled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service