653896
3-(Boc-aminomethyl)piperidine
95%
Synonym(s):
(3-Piperidinylmethyl)-carbamic acid 1,1-dimethylethyl ester, (Piperidin-3-ylmethyl) carbamic acid tert-butyl ester, tert-Butyl (piperidin-3-ylmethyl) carbamate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C5H10NCH2NHCO2C4H9
CAS Number:
Molecular Weight:
214.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
form
solid
mp
60-70 °C (lit.)
functional group
amine
SMILES string
CC(C)(C)OC(=O)NCC1CCCNC1
InChI
1S/C11H22N2O2/c1-11(2,3)15-10(14)13-8-9-5-4-6-12-7-9/h9,12H,4-8H2,1-3H3,(H,13,14)
InChI key
KHPQHXGYYXYTDN-UHFFFAOYSA-N
Related Categories
Application
Reagent for:
- Preparation of 9-substituted phenanthrene-based tylophorine analogs
- Enzymatic acylation
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service