Skip to Content
Merck
All Photos(1)

Key Documents

641596

Sigma-Aldrich

Quinoline-d7

97 atom % D, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D7N
Molecular Weight:
136.20
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

97 atom % D

Assay

98% (CP)

bp

237 °C (lit.)

mp

-17--13 °C (lit.)

density

1.151 g/mL at 25 °C

mass shift

M+7

SMILES string

[2H]c1nc2c([2H])c([2H])c([2H])c([2H])c2c([2H])c1[2H]

InChI

1S/C9H7N/c1-2-6-9-8(4-1)5-3-7-10-9/h1-7H/i1D,2D,3D,4D,5D,6D,7D

InChI key

SMWDFEZZVXVKRB-GSNKEKJESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service