Skip to Content
Merck
All Photos(3)

Key Documents

638099

Sigma-Aldrich

CyJohnPhos

97%

Synonym(s):

2-(Dicyclohexylphosphino)biphenyl, (2-Biphenyl)dicyclohexylphosphine, Cyclohexyl JohnPhos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31P
CAS Number:
Molecular Weight:
350.48
Beilstein:
8440533
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-X Bond Formation

reagent type: ligand
reaction type: Hiyama Coupling

reagent type: ligand
reaction type: Methylations

reagent type: ligand
reaction type: Negishi Coupling

reagent type: ligand
reaction type: Oxidations

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

mp

102-106 °C (lit.)

functional group

phosphine

SMILES string

C1CCC(CC1)P(C2CCCCC2)c3ccccc3-c4ccccc4

InChI

1S/C24H31P/c1-4-12-20(13-5-1)23-18-10-11-19-24(23)25(21-14-6-2-7-15-21)22-16-8-3-9-17-22/h1,4-5,10-13,18-19,21-22H,2-3,6-9,14-17H2

InChI key

LCSNDSFWVKMJCT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CyJohnPhos [(2-Biphenyl)dicyclohexylphosphine] is an air-stable, bulky and electron-rich monodentate biarylphosphine ligand developed by the Buchwald group to enhance the reactivity of palladium catalysis during cross-coupling reactions.

Learn more about Buchwald Phosphine Ligands

Application

CyJohnPhos may be used in combination with a nickel catalyst for the Suzuki-Miyaura arylation of tertiary benzylic acetates with aryl boroxines to produce diaryl and triaryl quaternary stereocenters.
Ligand employed in the palladium-catalysed synthesis of 1,3,5-tris(2′-aminophenyl)benzene, a core building block for potential ionic receptors.
Ligand for amination of aryl halides and triflates.

Legal Information

Usage subject to US Patents 6307087 and 6395916.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synlett, 2027-2027 (2006)
Stereospecific Cross Couplings To Set Benzylic, All-Carbon Quaternary Stereocenters in High Enantiopurity.
Zhou Q, et al.
Journal of the American Chemical Society, 138(37), 12057-12060 (2016)
Daichi Kakizoe et al.
Dalton transactions (Cambridge, England : 2003), 46(43), 14804-14811 (2017-10-19)
We report the synthesis and photophysical properties of Cu(i) complexes with 1,10-phenanthroline (phen) and monodentate phosphine ligands. Single crystal X-ray structural analysis revealed that these have three-coordinated trigonal planar geometries. We also found that one of them, [Cu(phen)(Johnphos)]BF
David S Surry et al.
Angewandte Chemie (International ed. in English), 47(34), 6338-6361 (2008-07-30)
Palladium-catalyzed amination reactions of aryl halides have undergone rapid development in the last 12 years, largely driven by the implementation of new classes of ligands. Biaryl phosphanes have proven to provide especially active catalysts in this context. This Review discusses
David S Surry et al.
Chemical science, 2(1), 27-50 (2011-01-01)
Dialkylbiaryl phosphines are a valuable class of ligand for Pd-catalyzed amination reactions and have been applied in a range of contexts. This review attempts to aid the reader in the selection of the best choice of reaction conditions and ligand

Articles

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

Buchwald and coworkers develop versatile phosphine ligands for Pd-catalyzed C–N bond formation; enhancing synthetic reactions for 20 years.

See All

Related Content

Explore reliable, premium grade catalysis materials for your pharma or industrial project. Specialty chemicals and formulations are available in bulk quantities and volumes from a few grams to multi-metric tons with complete documentation to simplify your leap from development to commercialization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service