Skip to Content
Merck
All Photos(2)

Key Documents

634638

Sigma-Aldrich

Yttrium aluminum oxide

nanopowder, <150 nm particle size (TEM), 99% trace metals basis

Synonym(s):

Aluminum yttrium oxide, YAG, Yttrium aluminum garnet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y3Al5O12
CAS Number:
Molecular Weight:
593.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99% trace metals basis

form

nanopowder

particle size

<150 nm (TEM)

density

4.56 g/mL at 25 °C (lit.)

SMILES string

O=[Al]O[Al]=O.O=[Al]O[Al]=O.O=[Al]O[Y]=O.O=[Y]O[Y]=O

InChI

1S/5Al.12O.3Y

InChI key

ITMSSWCUCPDVED-UHFFFAOYSA-N

Features and Benefits

Lanthanide doped YAG is the prime optical material in modern solid state lasers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesca Fumagalli et al.
Journal of the American Heart Association, 9(24), e016494-e016494 (2020-12-09)
Background Ventilation with the noble gas argon (Ar) has shown neuroprotective and cardioprotective properties in different in vitro and in vivo models. Hence, the neuroprotective effects of Ar were investigated in a severe, preclinically relevant porcine model of cardiac arrest.
Julia Wattacheril et al.
PloS one, 8(2), e57165-e57165 (2013-03-02)
Nonalcoholic fatty liver disease (NAFLD) occurs frequently in a setting of obesity, dyslipidemia and insulin resistance, but the etiology of the disease, particularly the events favoring progression to nonalcoholic steatohepatitis (NASH) as opposed to simple steatosis (SS), are not fully
Sumit Kumar Panja et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 20(7), 936-940 (2019-03-02)
Several recent studies of hydroxyl-functionalized ionic liquids (ILs) have shown that cation-cation interactions can be dominating these materials at the molecular level when the anion involved is weakly interacting. The hydrogen bonds between the like ions led to the formation
T André et al.
Nature communications, 9(1), 1334-1334 (2018-04-08)
With gigaelectron-volts per centimetre energy gains and femtosecond electron beams, laser wakefield acceleration (LWFA) is a promising candidate for applications, such as ultrafast electron diffraction, multistaged colliders and radiation sources (betatron, compton, undulator, free electron laser). However, for some of
Luciano Quaranta et al.
Advances in therapy, 36(9), 2506-2514 (2019-07-14)
The CoQun® study is a multicenter, controlled trial aimed to evaluate the neuroprotective effects of Coqun®, an ophthalmic solution of Coenzyme q10 (CoQ10) and Vitamin E (VitE), in patients affected by primary open-angle glaucoma (POAG). Pre-clinical studies and small non-controlled

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service