All Photos(1)
About This Item
Linear Formula:
HOCH2CH2N(CD3)3Br
CAS Number:
Molecular Weight:
193.13
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
98 atom % D
form
solid
mass shift
M+9
SMILES string
[Br-].[2H]C([2H])([2H])[N+](CCO)(C([2H])([2H])[2H])C([2H])([2H])[2H]
InChI
1S/C5H14NO.BrH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1/i1D3,2D3,3D3;
InChI key
JJCWKVUUIFLXNZ-KYRNGWDOSA-M
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service