615307
3-Ethyl-3-pentanol-OD
98 atom % D
Synonym(s):
3-Ethyl-3-pentan(ol-d)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5)3C-OD
Molecular Weight:
117.21
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
Recommended Products
isotopic purity
98 atom % D
density
0.831 g/mL at 25 °C
mass shift
M+1
SMILES string
[2H]OC(CC)(CC)CC
InChI
1S/C7H16O/c1-4-7(8,5-2)6-3/h8H,4-6H2,1-3H3/i8D
InChI key
XKIRHOWVQWCYBT-BNEYPBHNSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
3 - Flammable Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service