Skip to Content
Merck
All Photos(1)

Key Documents

608114

Sigma-Aldrich

L-Proline-13C5,15N

98 atom % 15N, 98 atom % 13C, 95% (CP)

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid-13C5,15N, L-Proline-ul-13C,15N, 13C and 15N Labeled L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C5H915NO2
CAS Number:
Molecular Weight:
121.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % 13C
98 atom % 15N

Assay

95% (CP)

form

powder

optical activity

[α]25/D -86.0°, c = 2 in H2O

technique(s)

bio NMR: suitable

mp

228 °C (dec.) (lit.)

mass shift

M+6

storage temp.

room temp

SMILES string

O[13C](=O)[13C@@H]1[13CH2][13CH2][13CH2][15NH]1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1

InChI key

ONIBWKKTOPOVIA-XAFSXMPTSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles N Zawatsky et al.
Frontiers in endocrinology, 12, 744857-744857 (2021-10-16)
Scleroderma, or systemic sclerosis, is a multi-organ connective tissue disease resulting in fibrosis of the skin, heart, and lungs with no effective treatment. Endocannabinoids acting via cannabinoid-1 receptors (CB1R) and increased activity of inducible NO synthase (iNOS) promote tissue fibrosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service