Skip to Content
Merck
All Photos(3)

Key Documents

594709

Sigma-Aldrich

2-Bromo-3′-hydroxyacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7BrO2
CAS Number:
Molecular Weight:
215.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

70-74 °C (lit.)

functional group

bromo
ketone

SMILES string

Oc1cccc(c1)C(=O)CBr

InChI

1S/C8H7BrO2/c9-5-8(11)6-2-1-3-7(10)4-6/h1-4,10H,5H2

InChI key

IEPSGFQQGKPTPM-UHFFFAOYSA-N

Related Categories

Application

Reactant involved in the synthesis of:
  • Pyrrole-3-carboxylic acids
  • N-aryl-N-thiazolyl compounds via the Hantzsch reaction
  • Disubstituted oxadiazolylindole derivatives with antiinflammatory, analgesic and nitric oxide releasing activity
  • Anilino-aryl-thiazoles with inhibitory activity toward valosin-containing proteins
  • Tyrosine kinase erythropoietin inhibitors
  • E. coli methionine aminopeptidase inhibitors

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service