Skip to Content
Merck
All Photos(1)

Key Documents

578576

Sigma-Aldrich

1,2-Epoxy-3-phenoxypropane

technical grade

Synonym(s):

2,3-Epoxypropyl phenyl ether, Glycidyl phenyl ether, Phenyl glycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
Beilstein:
2744
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

vapor density

5.2 (vs air)

vapor pressure

0.03 mmHg ( 20 °C)

refractive index

n20/D 1.53 (lit.)
n20/D 1.530 (lit.)

bp

245 °C (lit.)

mp

3.5 °C (lit.)

density

1.109 g/mL at 25 °C (lit.)

SMILES string

C1OC1COc2ccccc2

InChI

1S/C9H10O2/c1-2-4-8(5-3-1)10-6-9-7-11-9/h1-5,9H,6-7H2

InChI key

FQYUMYWMJTYZTK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1B - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Van den Eeckhout et al.
Journal of chromatography, 504(1), 113-128 (1990-03-30)
Thymidine and 2'-deoxyadenosine were reacted with phenyl glycidyl ether in order to study the formation of the corresponding 2'-deoxynucleoside adducts. Separation methods were elaborated using either reversed-phase high-performance liquid chromatography with photodiode-array detection, or centrifugal circular thin-layer chromatography. The adducts
Pablo Taboada et al.
Langmuir : the ACS journal of surfaces and colloids, 22(18), 7465-7470 (2006-08-23)
The aim of this study was to define a block copolymer micellar system with a high solubilization capacity for poorly soluble aromatic drugs. Ethylene oxide and phenyl glycidyl ether were sequentially polymerized to form the diblock copolymer G5E67 (G =
D L Deforce et al.
Analytical biochemistry, 258(2), 331-338 (1998-05-08)
The in vitro adduct formation between phenyl glycidyl ether (PGE) and calf thymus DNA was investigated. Agarose slab gel electrophoresis of DNA incubated with PGE revealed that nearly all high-molecular-weight species were degraded after 10 h of incubation. After DNA
The inhalation toxicity of phenylglycidyl ether: reproduction, mutagenic, teratogenic, and cytogenic studies.
J B Terrill et al.
Toxicology and applied pharmacology, 64(2), 204-212 (1982-06-30)
Ida B Niklasson et al.
Chemical research in toxicology, 24(4), 542-548 (2011-03-05)
Epoxy resins are among the most common causes of occupational contact dermatitis. They are normally used in so-called epoxy resin systems (ERS). These commercial products are combinations of epoxy resins, curing agents, modifiers, and reactive diluents. The most frequently used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service