Skip to Content
Merck
All Photos(1)

Key Documents

571032

Sigma-Aldrich

Poly[9,9-bis-(2-ethylhexyl)-9H-fluorene-2,7-diyl]

light-emitting λem 409 nm (in chloroform)

Synonym(s):

Poly[9,9-di-(2′-ethylhexyl)fluorenyl-2,7-diyl]

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

transition temp

Tg (DSC) 45 °C (onset)

solubility

THF: soluble
chloroform: soluble
toluene: soluble
xylene: soluble

λmax

374 nm

fluorescence

λex 362 nm; λem 409 nm in chloroform

SMILES string

C1(c2c(cccc2)c3c1cccc3)(CC(CCCC)CC)CC(CCCC)CC

InChI key

YJUVFJKHTNWDNU-UHFFFAOYSA-N

General description

Light-emitting, conjugated polymer exhibiting blue photo- and electroluminescence.
Poly[9,9-bis-(2-ethylhexyl)-9H-fluorene-2,7-diyl] (PFO) is a polyfluorene based conductive liquid crystalline material that can be used as a light emitting material. It forms a semiconducting layer to enhance the electroluminescence (EL) and performance of the photonic and optoelectronic devices.[1][2]

Application

PFO forms a π-conjugating polymeric system that can produce blue EL devices which include organic light emitting diodes (OLEDs), optical fibers and molecular electronic devices.[3][4][5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blue polarized electroluminescence from a liquid crystalline polyfluorene
Grell M, et al.
Advanced Materials, 11(8), 671-675 (1999)
Polymer Light-emitting Diodes Based on End-capped Poly [9, 9-di-(2′ -ethylhexyl) fluorenyl-2, 7-diyl]
Zhang Q
Chin. J. Chem., 28(8), 1482-1486 (2010)
Amplified spontaneous emission spectra of poly (9, 9-dioctylfluorenyl-2, 7-diyl) under pulsed laser excitation
Ibnaouf KH
Synthetic Metals, 209, 534-543 (2015)
Design and performance of active coupler for plastic optical fibres
Zubia J, et al.
Electronics Letters, 38(2), 65-67 (2002)
A Blue-Light-Emitting Polymer with a Rigid Backbone for Enhanced Color Stability
Park SH, et al.
Advances in Functional Materials, 17(16), 3063-3068 (2007)

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service