Skip to Content
Merck
All Photos(1)

Key Documents

553344

Sigma-Aldrich

Isopropyl isocyanide

97%

Synonym(s):

2-Isocyanopropane, 2-Propyl isonitrile, Isopropyl isonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNC
CAS Number:
Molecular Weight:
69.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

refractive index

n20/D 1.371 (lit.)

bp

75 °C (lit.)

density

0.733 g/mL at 25 °C (lit.)

functional group

amine
isonitrile

storage temp.

2-8°C

SMILES string

CC(C)[N+]#[C-]

InChI

1S/C4H7N/c1-4(2)5-3/h4H,1-2H3

InChI key

MJZUMMKYWBNKIP-UHFFFAOYSA-N

General description

Isopropyl isocyanide is an alkyl isocyanide.

Application

Isopropyl isocyanide may be used in the preparation of:
  • Ugi ligand A2C11I1, which is employed in the solid-phase Ugi synthesis.[1]
  • Pentafluorophenyl (PFP) functional monomers, via Passerini reaction.[2]
  • Dinuclear copper complex [Cu2(μ-PiPr2bipy)2{μ-CNCH(CH3)2}](PF6)2 [PiPr2bipy = 6-(diisopropylphosphanyl)-2,2′-bipyridine].[3]
  • (3E)-(Imino)thiaisoindoline 1,1-dioxide derivative.[4]
  • N,N′-diisopropylcarbodiimide (DIC) analog via reaction with propan-2-amine.[5]
  • 1-t-butyl-2-isopropyldiaziridinone via reaction with 2-methyl-2-nitrosopropane[6]
  • isopropyl isocyanate via reaction with ozone[7]
  • 1-phthalimidoazetidines via [1+3]cycloaddition reaction with 1-pththalimidoaziridines[8]

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.1 °F - closed cup

Flash Point(C)

20.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[1+ 3] Cycloadditions of isocyanides to azomethine ylides; synthesis and properties of 1-phthalimidoazetidines
Charrier J, et al.
The Journal of Organic Chemistry, 48(4), 481-486 (1983)
Reaction of Alkyl Isocyanides with Ozone. A New Isocyanate Synthesis1
Feuer H, et al.
The Journal of Organic Chemistry, 23(8), 1107-1109 (1958)
A synthetic Protein G adsorbent based on the multi-component Ugi reaction for the purification of mammalian immunoglobulins.
Qian J, et al.
Journal of Chromatography. B, Biomedical Applications, 898, 15-23 (2012)
Palladium-Catalyzed C C Coupling of Aryl Halides with Isocyanides: An Alternative Method for the Stereoselective Synthesis of (3E)-(Imino) isoindolin-1-ones and (3E)-(Imino) thiaisoindoline 1, 1-Dioxides.
Liu B, et al.
Advanced Synthesis & Catalysis, 354(11-12), 2288-2300 (2012)
A Series of Dinuclear Copper Complexes Bridged by Phosphanylbipyridine Ligands: Synthesis, Structural Characterization and Electrochemistry.
Lilio AM, et al.
European Journal of Organic Chemistry, 22-23, 4016-4023 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service