Skip to Content
Merck
All Photos(3)

Key Documents

538701

Sigma-Aldrich

2-Ethylhexanoic acid

≥99%

Synonym(s):

2-Ethylcaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CO2H
CAS Number:
Molecular Weight:
144.21
Beilstein:
1750468
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.98 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
10 mmHg ( 115 °C)

Assay

≥99%

autoignition temp.

699 °F

expl. lim.

1.04 %, 135 °F
8.64 %, 188 °F

refractive index

n20/D 1.425 (lit.)

bp

228 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCC(CC)C(O)=O

InChI

1S/C8H16O2/c1-3-5-6-7(4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

OBETXYAYXDNJHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexanoic acid (EHXA, 2-EHA) is an industrially important aliphatic carboxylic acid. It is widely employed as a stabilizer and a wood preservative.
It has various industrial applications, such as:
  • coolant in automotives
  • synthetic lubricant
  • wetting agent
  • co-solvent
  • drying of paints
  • defoaming agent in pesticides

Application

  • A review of the environmental fate and aquatic effects of a series of C4 and C8 oxo-process chemicals.: This review assesses the environmental impact of C4 and C8 oxo-process chemicals, including 2-Ethylhexanoic acid, detailing their degradation, persistence, and effects on aquatic ecosystems (Staples, 2001).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The developmental toxicity of 2-ethylhexanoic acid in Wistar rats.
Pennanen S, et al.
Fundamental and Applied Toxicology, 19(4), 505-511 (1992)
Synthetic applications of 2-ethylhexanoic acid derived reagents.
Raju R and Prasad K.
Tetrahedron, 68(5), 1341-1349 (2012)
K J French et al.
Biochemistry, 40(32), 9532-9538 (2001-10-05)
This study examines the ability of P450cam to catalyze the formation of 2-ethylhexanoic acid from 2-ethylhexanol relative to its activity on the natural substrate camphor. As is the case for camphor, the P450cam exhibits stereoselectivity for binding (R)- and (S)-2-ethylhexanol.
Shashank Mishra et al.
Chemical Society reviews, 36(11), 1770-1787 (2008-01-25)
This critical review deals with the chemistry and applications of metal alkanoates with medium size (C5 to C12) carbon chain length. A particular emphasis is given to metal 2-ethylhexanoates, which find wide applications as metal-organic precursors in materials science, as
R Gaudin et al.
International archives of occupational and environmental health, 84(5), 523-531 (2010-08-31)
The aim of this study was to assess, by biological monitoring, workers' exposure to di(2-ethylhexyl) phthalate (DEHP) in the flexible-PVC industry in France to provide additional occupational exposure data, which are particularly scarce. Over 5 days of pre-and post-shift sampling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service