Skip to Content
Merck
All Photos(1)

Documents

531162

Sigma-Aldrich

Poly(D,L-lactide)

inherent viscosity 0.55-0.75 dL/g (lit.)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mw 75,000-120,000

degradation timeframe

12-16 months

inherent viscosity

0.55-0.75 dL/g(lit.)

mp

262 °C (lit.)

transition temp

Tg 32.9 °C

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Physical properties

Biocompatible, biodegradable polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service