Skip to Content
Merck
All Photos(1)

Key Documents

528870

Sigma-Aldrich

5-Bromo-2-fluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)CHO
CAS Number:
Molecular Weight:
203.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.57 (lit.)

bp

230 °C (lit.)

density

1.71 g/mL at 25 °C (lit.)

functional group

aldehyde
bromo
fluoro

storage temp.

2-8°C

SMILES string

Fc1ccc(Br)cc1C=O

InChI

1S/C7H4BrFO/c8-6-1-2-7(9)5(3-6)4-10/h1-4H

InChI key

MMFGGDVQLQQQRX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.9 °F - closed cup

Flash Point(C)

109.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 5-cyanoindazole and 1-methyl and 1-aryl-5-cyanoindazoles.
Halley F and Sava X.
Synthetic Communications, 27(7), 1199-1207 (1997)
Christian Ruzié et al.
The Journal of organic chemistry, 78(15), 7741-7748 (2013-07-16)
The synthesis of 1,6-, 2,7-, 3,8-, and 4,9-isomers of dibromo- and didodecyl[1]benzothieno[3,2-b][1]benzothiophenes, via the stilbene pathway, is described. Starting from the synthesis of bromo-2-(methylthio)benzaldehydes, a series of functionalization, McMurry coupling, and finalising cyclization reactions were explored. The stereochemistry of the
High throughput synthesis of diverse 2, 5-disubstituted indoles using titanium carbenoids bearing boronate functionality.
Main CA, et al.
Tetrahedron, 64(5), 901-914 (2008)
Keith W Woods et al.
Bioorganic & medicinal chemistry, 14(20), 6832-6846 (2006-07-18)
A series of heteroaryl-pyridine containing inhibitors of Akt are reported. The synthesis and structure-activity relationships are discussed, leading to the discovery of a indazole-pyridine analogue (K(i)=0.16 nM). These compounds bind in the ATP binding site, are potent, ATP competitive, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service