Skip to Content
Merck
All Photos(3)

Key Documents

526339

Sigma-Aldrich

3-(Benzyloxy)phenylboronic acid

≥95%

Synonym(s):

3-Benzyloxybenzeneboronic acid, 3-Phenylmethyloxyphenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4B(OH)2
CAS Number:
Molecular Weight:
228.05
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

125-130 °C (lit.)

functional group

phenyl

SMILES string

OB(O)c1cccc(OCc2ccccc2)c1

InChI

1S/C13H13BO3/c15-14(16)12-7-4-8-13(9-12)17-10-11-5-2-1-3-6-11/h1-9,15-16H,10H2

InChI key

WIJNYNBSPQMJGO-UHFFFAOYSA-N

General description

Contains varying amounts of anhydride.

Application

Reactant for:
  • Microwave Suzuki-Miyaura coupling
  • Preparation of palladium-based fluoride-derived electrophilic fluorination reagent for PET imaging agents
  • Synthesis of substituted isoindolines via palladium-catalyzed cascade reaction
  • Ruthenium-catalyzed hydrogenation
  • Suzuki coupling reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service