Skip to Content
Merck
All Photos(2)

Key Documents

520780

Sigma-Aldrich

Platinum black

greener alternative

≥99.9% trace metals basis

Synonym(s):

Platinum element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: platinum

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

25-34 m2/g

greener alternative category

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Platinum black (fuel cell grade) can be electroplated on implantable microelectrode arrays (MEAs) to improve the impedence of the device.[1] It can be used as a microelectrode for the formation of a microfluidic electrochemical sensor for glucose sensing applications.[2] It can also be used as a cathode material for the production of hydrogen and polymer electrolyte fuel cells.[3][4]

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Progress in non-precious metal oxide-based cathode for polymer electrolyte fuel cells
Ishihara A, et al.
Electrochimica Acta, 55(27), 8005-8012 (2010)
Revealing the semiconductor-catalyst interface in buried platinum black silicon photocathodes
Aguiar JA, et al.
Journal of Material Chemistry A, 4(21), 8123-8129 (2016)
A droplet-based microfluidic electrochemical sensor using platinum-black microelectrode and its application in high sensitive glucose sensing
Gu S, et al.
Biosensors And Bioelectronics, 55, 106-112 (2014)
Improving impedance of implantable microwire multi-electrode arrays by ultrasonic electroplating of durable platinum black
Arcot Desai S, et al.
Frontiers in Neuroengineering, 3, 5-5 (2010)
Hiroaki Akamatsu et al.
Japanese journal of clinical oncology, 43(6), 664-668 (2013-04-16)
Interstitial lung disease associated with gefitinib is a critical adverse reaction. When geftinib was administered to EGFR-unknown patients, the interstitial lung disease incidence rate was approximately 3-4% in Japan, and usually occurs during the first 4 weeks of treatment. However

Articles

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service