516244
Calcium iodide
AnhydroBeads™, −10 mesh, 99.95% trace metals basis
Synonym(s):
Calcium diiodide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CaI2
CAS Number:
Molecular Weight:
293.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.95% trace metals basis
Recommended Products
product line
AnhydroBeads™
Quality Level
Assay
99.95% trace metals basis
impurities
≤550.0 ppm Trace Metal Analysis
particle size
−10 mesh
mp
779 °C (lit.)
SMILES string
I[Ca]I
InChI
1S/Ca.2HI/h;2*1H/q+2;;/p-2
InChI key
UNMYWSMUMWPJLR-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Calcium iodide can be used:
- As a passivator of CH3NH3PbI3(MAPbI3) perovskite films to achieve high efficiency and long-term stability.
- As a dopant to prepare 3D γ-CsPbI3 perovskite solar cells, to improve their stability and power conversion efficiency.
- As a precursor to synthesize a series of heteroleptic calcium formamidinate derivatives.
- As a catalyst for photooxidative oxylactonization of oxocarboxylic acids.
Legal Information
AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC
accessory
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Calcium iodide catalyzed photooxidative oxylactonization of oxocarboxylic acids using molecular oxygen as terminal oxidant
Norihiro Tada, et al
Tetrahedron Letters, 54, 256-258 (2013)
Preparation of Tortuous 3D gamma-CsPbI3 Films at Low Temperature by CaI2 as Dopant for Highly Efficient Perovskite Solar Cells
Hang Zhao, et al.
Advanced Functional Materials
, 29, 1808986-1808986 (2019)
CaI2:a more effective passivator of perovskite films than PbI2 for high efficiency and long-term stability of perovskite solar cells
Chuanliang Chen, et al.
Journal of Materials Chemistry, 6, 7903-7912 (2018)
Calcium formamidinate derivatives by hydride insertion of carbodiimides
Joshua Dyall, et al.
Dalton Transactions, 48, 4248-4254 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service