Skip to Content
Merck
All Photos(1)

Key Documents

50640

Sigma-Aldrich

Glycolic acid nitrile solution

~70% in H2O

Synonym(s):

Formaldehyde cyanohydrin, Hydroxyacetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CN
CAS Number:
Molecular Weight:
57.05
Beilstein:
605328
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

contains

~0.5% phosphoric acid as stabilizer

Quality Level

concentration

~70% in H2O

refractive index

n20/D 1.389

density

1.076 g/mL at 20 °C

functional group

hydroxyl
nitrile

SMILES string

OCC#N

InChI

1S/C2H3NO/c3-1-2-4/h4H,2H2

InChI key

LTYRAPJYLUPLCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The product is a 70% solution of glycolic acid nitrile in water. Glycolic acid nitrile, also known as hydroxyacetonitrile is the simplest cyanohydrin. The microwave spectra of hydroxyacetonitrile have been recorded in gas-phase and its rotational constants have been calculated. Ice containing formaldehyde and hydrogen cyanide react under astrophysical-like conditions to form hydroxyacetonitrile.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroxyacetonitrile (HOCH2CN) formation in astrophysical conditions. competition with the aminomethanol, a glycine precursor.
Danger G, et al.
The Astrophysical Journal, 756(1), 11-11 (2012)
Microwave and Quantum-Chemical Study of Conformational Properties and Intramolecular Hydrogen Bonding of 2-Hydroxy-3-Butynenitrile (HC=CCH(OH)C=N).
M?llendal H, et al.
The Journal of Physical Chemistry A, 119(4), 634-640 (2015)
Formation of hydroxyacetonitrile (HOCH2CN) and polyoxymethylene (POM)-derivatives in comets from formaldehyde (CH2O) and hydrogen cyanide (HCN) activated by water.
Danger G, et al.
Physical Chemistry Chemical Physics, 16(8), 3360-3370 (2014)
Rotational isomerism and barriers to internal rotation in hydroxyacetonitrile from microwave spectroscopy.
Cazzoli G, et al.
J. Chem. Soc., Faraday II, 69, 569-578 (1973)
Evidence for the microsomal metabolism of glycolonitrile.
J J Freeman et al.
Biochemical pharmacology, 36(1), 184-187 (1987-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service