All Photos(1)
About This Item
Linear Formula:
H215N(CH2)4CH(NH2)CO2H · 2HCl
CAS Number:
Molecular Weight:
220.10
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Recommended Products
isotopic purity
98 atom % 15N
Assay
99% (CP)
form
solid
mp
190 °C (lit.)
mass shift
M+1
SMILES string
Cl.Cl.NC(CCCC[15NH2])C(O)=O
InChI
1S/C6H14N2O2.2ClH/c7-4-2-1-3-5(8)6(9)10;;/h5H,1-4,7-8H2,(H,9,10);2*1H/i7+1;;
InChI key
JBBURJFZIMRPCZ-ADOWEKEWSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service