Skip to Content
Merck
All Photos(1)

Key Documents

489336

Sigma-Aldrich

Ethanol-1,1,2,2,2-d5

99.5 atom % D

Synonym(s):

Ethyl-d5 alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2OH
CAS Number:
Molecular Weight:
51.10
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99.5 atom % D

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.358 (lit.)

bp

78 °C (lit.)

density

0.871 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])O

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1D3,2D2

InChI key

LFQSCWFLJHTTHZ-ZBJDZAJPSA-N

General description

Ethanol-1,1,2,2,2-d5 (ethyl-d5 alcohol, CD3CD2OH) is deuterated ethyl alcohol. The nuclear magnetic relaxation rates of 13C in 13CCl4 and CD3CD2OH solvent mixture has been determined. The quantification of ethyl-d5 alcohol along with ethyl alcohol in serum by stable isotope gas chromatography-mass spectrometry technique has been reported. It plays a role as a tracer in the time-of-flight secondary ion mass spectrometry (TOF-SIMS) tracer method to assess the boundary lubricating behaviors of boundary lubricating films.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous measurement of ethanol and ethyl-d5 alcohol by stable isotope gas chromatography-mass spectrometry.
Dean RA, et al.
Clinical Chemistry, 42(3), 367-372 (1996)
Joon-Bae Lee et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-28)
Methanol is metabolized in the body to highly toxic formaldehyde and formate when consumed accidentally. Methanol has been typically analyzed with gas chromatography-flame ionization detector (GC-FID). However, its retention time may overlap with other volatile compounds and lead to confusion.
Relative molecular orientation in ethanol-CCl4 mixtures as determined by nuclear magnetic relaxation mesurements.
Langer H, et al.
Chemical Physics Letters, 23(3), 417-421 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service