Skip to Content
Merck
All Photos(1)

Documents

488577

Sigma-Aldrich

Cholesterol-2,2,3,4,4,6-d6

97 atom % D, 98% (CP)

Synonym(s):

3β-Hydroxy-5-cholestene-2,2,3,4,4,5-d6, 5-Cholesten-3β-ol-2,2,3,4,4,5-d6, Deuterated cholesterol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D6H40O
CAS Number:
Molecular Weight:
392.69
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

97 atom % D

Assay

98% (CP)

optical activity

[α]20/D -40°, c = 2 in chloroform

technique(s)

mass spectrometry (MS): suitable

bp

360 °C (lit.)

mp

147-149 °C (lit.)

density

1.088 g/mL at 25 °C

mass shift

M+6

storage temp.

−20°C

SMILES string

[2H]C1=C2C([2H])([2H])[C@@]([2H])(O)C([2H])([2H])C[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3C1)[C@H](C)CCCC(C)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i9D,13D2,17D2,21D

InChI key

HVYWMOMLDIMFJA-HHPLMCQASA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Passoni et al.
ACS chemical neuroscience, 11(3), 367-372 (2019-12-21)
The current pharmacological treatment of Huntington's disease (HD) is palliative, and therapies to restore functions in patients are needed. One of the pathways affected in HD involves brain cholesterol (Chol) synthesis, which is essential for optimal synaptic transmission. Recently, it
Matteo Ludovici et al.
Scientific reports, 8(1), 11500-11500 (2018-08-02)
The skin surface lipids (SSL) result from the blending of sebaceous and epidermal lipids, which derive from the sebaceous gland (SG) secretion and the permeability barrier of the stratum corneum (SC), respectively. In humans, the composition of the SSL is
Yasuyo Okada et al.
Carbohydrate research, 455, 54-61 (2017-11-25)
We aimed to investigate whether 6-O-α-maltosyl-β-cyclodextrin (Mal-βCD) is incorporated into cells and lysosomes during the release of unesterified cholesterol in Npc1-deficient Chinese hamster ovary (CHO) cells (Npc1 KO cells) and CHO-JP17 cells (JP17 cells). Internalization of Mal-βCD in cells and lysosomes and
Wenxu Zhou et al.
Journal of lipid research, 61(2), 192-204 (2019-09-25)
Primitive sterol evolution plays an important role in fossil record interpretation and offers potential therapeutic avenues for human disease resulting from nematode infections. Recognizing that C4-methyl stenol products [8(14)-lophenol] can be synthesized in bacteria while C4-methyl stanol products (dinosterol) can
David Wheeler et al.
Brain : a journal of neurology, 131(Pt 11), 3092-3102 (2008-09-06)
Maintaining the appropriate complement and content of lipids in cellular membranes is critical for normal neural function. Accumulating evidence suggests that even subtle perturbations in the lipid content of neurons and myelin can disrupt their function and may contribute to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service