Skip to Content
Merck
All Photos(1)

Key Documents

487406

Sigma-Aldrich

4-Methoxybenzyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2NCO
CAS Number:
Molecular Weight:
163.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

refractive index

n20/D 1.433 (lit.)

bp

60-66 °C/0.6 mmHg (lit.)

density

1.143 g/mL at 25 °C (lit.)

functional group

amine
isocyanate

storage temp.

2-8°C

SMILES string

COc1ccc(CN=C=O)cc1

InChI

1S/C9H9NO2/c1-12-9-4-2-8(3-5-9)6-10-7-11/h2-5H,6H2,1H3

InChI key

QRBHVARIMDDOOV-UHFFFAOYSA-N

General description

4-Methoxybenzyl isocyanate (PMBNCO), also known as p-methoxybenzyl isocyanate, is an organic building block containing an isocyanate group. Its enthalpy of vaporization at boiling point has been reported.

Application

4-Methoxybenzyl isocyanate (p-Methoxybenzyl isocyanate) may be used in the synthesis of 1-(4-methoxybenzyl)-3-{2-[(4-nitrophenyl)amino]ethyl}urea and 1-(4-methoxybenzyl)-3-{2-[(5-nitropyridin-2-yl)amino]ethyl} urea.
It may be used in the multi-step synthesis of:
  • diimidazodiazepine containing 5:7:5-fused heterocycle
  • 2-deoxy-2-ureido-D-ribo-hexulofuranosonamide
  • bicyclic guanidine core of batzelladine A

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis and herbicidal activity of opened hydantoin-ring derivatives of hydantocidin.
Hiromi S, et al.
Bioscience, Biotechnology, and Biochemistry, 60(7), 1198-1200 (1996)
Tomasz M Wróbel et al.
Journal of enzyme inhibition and medicinal chemistry, 31(4), 608-618 (2015-06-27)
A series of urea derivatives bearing nitroaryl moiety has been synthesized and assayed for their potential antiproliferative activities. Some of the tested compounds displayed activity in RK33 laryngeal cancer cells and TE671 rhabdomyosarcoma cells while being generally less toxic to
Raj Kumar et al.
Organic letters, 10(20), 4681-4684 (2008-09-26)
The first synthesis of a novel 5:7:5-fused heterocyclic ring system, a diimidazodiazepine, is reported. The propensity of the ring system to undergo facile, acid-catalyzed nucleophilic addition reactions by neutral carbon and nitrogen nucleophiles has been explored. The ring system has
Michael A Arnold et al.
Journal of the American Chemical Society, 128(40), 13255-13260 (2006-10-05)
A diastereoselective [4 + 2]-annulation of vinyl carbodiimides with chiral N-alkyl imines has been developed to access the stereochemically rich polycyclic guanidine cores of the batzelladine alkaloids. Application of this strategy, together with additional key steps such as long-range directed
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 567-567 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service