Skip to Content
Merck
All Photos(3)

Documents

485926

Sigma-Aldrich

Sodium L-lactate-13C3 solution

45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral purity, HPLC)

Synonym(s):

L-Lactic acid-13C3 sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH313CH(OH)13CO2Na
CAS Number:
Molecular Weight:
115.04
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

description

Packaged as gram of solution 45-55%(w/w)

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)
≥98% (Chiral purity, HPLC)

concentration

45-55 % (w/w) in H2O

technique(s)

mass spectrometry (MS): suitable

mp

163-165 °C (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

[Na+].[13CH3][13C@H](O)[13C]([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i1+1,2+1,3+1;

InChI key

NGSFWBMYFKHRBD-USULVYIHSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium L-lactate-13C3 is an isotopic analogue of sodium L-lactate.

Application

Sodium L-lactate-13C3 can be used an internal standard for the quantitative determination of blood lactate and pyruvate using filter-paper dried blood spots followed by LC-MS/MS. It can also be used to study the metabolism of NAD redox metabolism or lactate levels, which may be help to develop novel immunotherapies in cancer and therapeutic immunosuppression.
Sodium L-lactate-13C3 has been used as an internal standard in the quantification of metabolites in murine tumor interstitial fluid using the LC/MS technique.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Wen Song et al.
Cell metabolism, 32(2), 188-202 (2020-07-02)
The coronavirus disease 2019 (COVID-19) pandemic presents an unprecedented threat to global public health. Herein, we utilized a combination of targeted and untargeted tandem mass spectrometry to analyze the plasma lipidome and metabolome in mild, moderate, and severe COVID-19 patients
Chih-Kuang Chuang et al.
Journal of chromatography. A, 1216(51), 8947-8952 (2009-11-17)
Lactic acidemia is commonly associated with severe diseases in pediatric patients. Quantitation of blood lactate and pyruvate is important for the diagnosis and clinical management. A liquid chromatography-tandem mass spectrometry (LC-MS/MS) method using dried blood spots (DBS) was developed and
M Testa et al.
Journal of ocular pharmacology, 3(3), 211-225 (1987-01-01)
Substances derived from biotransformation of non-steroid antiinflammatory drugs (NSAID) produced by patients responsive to the biological liquid oxidant activity (BLOA) test, have been shown to have anticataract activity. They are all aromatic alcohols with physico-chemical properties similar to benzyl alcohols
Isolation and Quantification of Metabolite Levels in Murine Tumor Interstitial Fluid by LC/MS
Sullivan MR, et al.
Bio-protocol, 9, e3427-e3427 (2019)
Minfeng Ying et al.
Redox biology, 46, 102065-102065 (2021-07-23)
Although glucose, through pentose phosphate pathway (PPP), is the main source to generate NADPH, solid tumors are often deprived of glucose, hence alternative metabolic pathways to maintain NADPH homeostasis in cancer cells are required. Here, we report that lactate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service