Skip to Content
Merck
All Photos(2)

Key Documents

485446

Sigma-Aldrich

Creatinine-(methyl-d3)

98 atom % D

Synonym(s):

4-Amidinosarcosine-d3 (methyl-d3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D3H4N3O
CAS Number:
Molecular Weight:
116.14
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

98 atom % D

Quality Level

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

255 °C (dec.) (lit.)

mass shift

M+3

SMILES string

[2H]C([2H])([2H])N1CC(=O)N=C1N

InChI

1S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)/i1D3

InChI key

DDRJAANPRJIHGJ-FIBGUPNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eulàlia Olesti et al.
Talanta, 192, 93-102 (2018-10-24)
Neurotransmitters are signaling molecules, playing key roles in neuronal communications in the brain. Drug induced changes in neurotransmitters and other brain metabolite concentration may be used to characterize drugs according to their targeted metabolomics profile. Here, we report the development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service