Skip to Content
Merck
All Photos(1)

Documents

47695

Sigma-Aldrich

Fmoc-Cys(Trt)-OH

≥95.0% (sum of enantiomers, HPLC)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-S-trityl-L-cysteine, Nα-Fmoc-S-trityl-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H31NO4S
CAS Number:
Molecular Weight:
585.71
Beilstein:
4221286
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥95.0% (sum of enantiomers, HPLC)

optical activity

[α]20/D +16.0±2°, c = 1% in THF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

170-173 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](CSC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C37H31NO4S/c39-35(40)34(38-36(41)42-24-33-31-22-12-10-20-29(31)30-21-11-13-23-32(30)33)25-43-37(26-14-4-1-5-15-26,27-16-6-2-7-17-27)28-18-8-3-9-19-28/h1-23,33-34H,24-25H2,(H,38,41)(H,39,40)/t34-/m0/s1

InChI key

KLBPUVPNPAJWHZ-UMSFTDKQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fmoc-Cys(Trt)-OH is an amino acid commonly used in the Fmoc solid-phase peptide synthesis.

Application

Fmoc-Cys(Trt)-OH is an N-terminal protected cysteine derivative used in peptide synthesis. Some of the examples are:
  • Synthesis of mono- and bi-functionalized platinum(IV) complexes to target angiogenic tumor vasculature.
  • Synthesis of proteins through native chemical ligation of peptide hydrazides as thioester surrogates via solid-phase synthesis.
  • Synthesis of glycoconjugates by conjugating reducing sugars to cysteine residues of peptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panchada Ch V Govindu et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 18(1), 198-207 (2018-11-14)
Avobenzone is the most widely used UVA filter in sunscreen lotion and it is prone to degradation in the presence of sunlight/UV radiation. To overcome the photo-instability of avobenzone, various photostabilizers have been used as additives, including antioxidants such as
Chemical synthesis of proteins using peptide hydrazides as thioester surrogates.
Zheng J S, et al.
Nature Protocols, 8(12), 2483-2483 (2013)
Parallel synthesis of structurally diverse aminobenzimidazole tethered sultams and benzothiazepinones
Sureshbabu D et al.
Tetrahedron Letters, 53, 6897-6900 (2012)
Conjugated platinum (IV)? peptide complexes for targeting angiogenic tumor vasculature.
Mukhopadhyay S, et al.
Bioconjugate Chemistry, 19(1), 39-49 (2007)
A double-click approach to the protecting group free synthesis of glycoconjugates.
Alexander S R, et al.
Organic & Biomolecular Chemistry, 16(8), 1258-1262 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service