Skip to Content
Merck
All Photos(1)

Key Documents

469319

Sigma-Aldrich

Poly(dimethylsiloxane)

viscosity 1.0 cSt (25 °C)

Synonym(s):

Octamethyltrisiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]2Si(CH3)2
CAS Number:
Molecular Weight:
236.53
Beilstein:
1753063
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

refractive index

n20/D 1.384 (lit.)

viscosity

1.0 cSt(25 °C)

bp

153 °C (lit.)

mp

−82 °C (lit.)

density

0.82 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

InChI

1S/C8H24O2Si3/c1-11(2,3)9-13(7,8)10-12(4,5)6/h1-8H3

InChI key

CXQXSVUQTKDNFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(dimethylsiloxane) (PDMS), also known as dimethicone, is an organosilicon compound that is widely used as a siloxane based elastomer. It is a hydrophobic material with attractive physical and chemical properties such as elasticity, optical transparency, tunable surface chemistry, and low electrical conductivity.[1][2][3]

Application

PDMS can be used in microfluidic devices,[4] nanolithography,[5] and other membrane based applications.[6][7]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - (External MSDS)

Flash Point(C)

29 °C - (External MSDS)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polylactide- poly (dimethylsiloxane)- polylactide triblock copolymers as multifunctional materials for nanolithographic applications
Rodwogin MD, et al.
ACS Nano, 4(2), 725-732 (2010)
Components for integrated poly (dimethylsiloxane) microfluidic systems
Ng JMK
Electrophoresis, 23(20), 3461-3473 (2002)
Soyoung Hong et al.
Polymers, 11(5) (2019-05-06)
The vascularization of three-dimensional (3D) tissue constructs is necessary for transporting nutrients and oxygen to the component cells. In this study, a vacuum forming method was applied to emboss a vascular pattern on an electrospun membrane so that guided vascular
Man Xu et al.
Journal of food science, 84(8), 2171-2180 (2019-07-18)
Amadori rearrangement product (ARP) derived from glutamic acid (Glu) and xylose (Xyl) was prepared by aqueous Maillard reaction. Subsequently, ion exchange chromatography, MS, and NMR were used for purification and identification, confirming that the molecular formula of ARP was C10
Chong In Shin et al.
Pharmaceutics, 11(7) (2019-08-03)
The stratum corneum is the outermost skin layer that obstructs the delivery of active ingredients found in cosmeceutical products. Chemical peels and microbeads have been used to overcome this layer, but these methods can cause side effects and are not

Articles

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Self-healing soft electronic materials enable autonomous repair, reducing waste and costs in electronic devices.

Related Content

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

We offer PDMS surface modification kit which allows for durable hydrophilization of various polydimethylsiloxane (PDMS) surfaces.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service