Skip to Content
Merck
All Photos(1)

Key Documents

468800

Sigma-Aldrich

4-Fluoro-D-phenylalanine hydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH(NH2)CO2H · HCl
CAS Number:
Molecular Weight:
219.64
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

4-Fluoro-D-phenylalanine hydrochloride, 99%

Assay

99%

form

powder

optical activity

[α]23/D +11°, c = 1 in H2O

color

white

mp

245 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cl.N[C@H](Cc1ccc(F)cc1)C(O)=O

InChI

1S/C9H10FNO2.ClH/c10-7-3-1-6(2-4-7)5-8(11)9(12)13;/h1-4,8H,5,11H2,(H,12,13);1H/t8-;/m1./s1

InChI key

ZDECBCKSULAIGP-DDWIOCJRSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service