All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
270-280 °C (dec.) (lit.)
storage temp.
2-8°C
SMILES string
O[C@H]1[C@@H](O)[C@@H](O)[C@@H](O)[C@H](O)[C@@H]1O
InChI
1S/C6H12O6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-12H/t1-,2-,3-,4-,5-,6+
InChI key
CDAISMWEOUEBRE-OQYPVSDDSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ileana-Andreea Ratiu et al.
Electrophoresis (2018-03-16)
Cyclitols are phytochemicals naturally occurring in plant material, which attracted an increasing interest due to multiple medicinal attributes, among which the most important are the antidiabetic, antioxidant, and anticancer properties. Due to their valuable properties, sugars are used in the
Ileana Andreea Ratiu et al.
Molecules (Basel, Switzerland), 25(1) (2019-12-22)
Honey is a natural sweetener, with an osmotic effect on microorganisms due to the increased sugar content and low amount of water. Cyclitols are minor constituents of honey. They play a defensive role in plants against unfavorable environmental conditions. Honey's
Sándor L Bekö et al.
IUCrJ, 1(Pt 1), 61-73 (2014-07-31)
Inositol, 1,2,3,4,5,6-hexahydroxycyclohexane, exists in nine stereoisomers with different crystal structures and melting points. In a previous paper on the relationship between the melting points of the inositols and the hydrogen-bonding patterns in their crystal structures [Simperler et al. (2006 ▶).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service