Skip to Content
Merck
All Photos(2)

Key Documents

466360

Sigma-Aldrich

Zinc iodide

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Diiodozinc, Zinc diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnI2
CAS Number:
Molecular Weight:
319.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.999% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: zinc

impurities

≤15.0 ppm Trace Metal Analysis

mp

445 °C (lit.)

density

4.74 g/mL at 25 °C (lit.)

SMILES string

I[Zn]I

InChI

1S/2HI.Zn/h2*1H;/q;;+2/p-2

InChI key

UAYWVJHJZHQCIE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y P Chau et al.
Histology and histopathology, 9(4), 649-656 (1994-10-01)
Cytochemical relationship between Golgi complex and dense-cored granules (DCGs) of small granule-containing (SGC) cells in rat superior cervical ganglia was examined in electron microscopy by zinc-iodide-osmium tetroxide (ZIO) method and by enzyme cytochemistry for thiamine pyrophosphatase (TPPase) and acid phosphatase
Yuko Nakamura et al.
Journal of endodontics, 31(10), 755-758 (2005-09-28)
The purpose of this study was to investigate the cytotoxicity of iontophoresis treatment using direct current (DC) with or without antibacterial agents. The following antibacterial agents were used: diamine silver fluoride (AgF); sodium fluoride (NaF); and iodine zinc iodide (JJZ).
T Müller
Histology and histopathology, 15(2), 493-498 (2000-05-16)
The nerve fiber distribution in the epidermis of the hairless rat skin was studied light microscopically by means of zinc iodide-osmium tetroxide staining. Two different morphological types of free nerve fiber endings could be detected: clusters of relatively thick nerve
Mohammad Saquib et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(24), 6041-6049 (2009-05-07)
Simple and efficient syntheses, catalysed by a mixed Lewis acid system (ZrCl(4)/ZnI(2)), of enantiomerically pure 2- and 2,3-disubstituted furan derivatives--including important synthons such as 3-iodofuran and 3-(hydroxymethyl)furan derivatives--from commercially available 3,4,6-tri-O-acetyl-D-glucal are described. The transformation is achieved through a synergistic
N Mayer-Gostan et al.
Cell and tissue research, 289(1), 53-61 (1997-07-01)
The saccular membranes of trout (Oncorhynchus mykiss) and turbot (Scophthalmus maximus) were examined to characterize specialized epithelial cells that might be responsible for ion exchange. The approach for localizing cell types was new for this tissue, as observations were made

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service