Skip to Content
Merck
All Photos(3)

Key Documents

457620

Sigma-Aldrich

Polyglycolide

inherent viscosity 1.4dL/g

Synonym(s):

PGA, Poly(glycolic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O3)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

solid

degradation timeframe

6-12 months

inherent viscosity

1.1-1.7 dL/g, 0.1 % (w/v) in hexafluoroisopropanol(25 °C)

transition temp

Tm 220-230 °C

solubility

hexafluoroisopropanol: soluble

density

1.53 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(O)=O

InChI

1S/C2H4O3/c3-1-2(4)5/h3H,1H2,(H,4,5)

InChI key

AEMRFAOFKBGASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biodegradable and biocompatible PGA could be potentially useful for bone regeneration. High molecular weight polyglycolide (PGA) can be prepared by ring opening polymerization of diglycolide in the presence of diphenyl bismuth bromide.

Application

Polyglycolide (PGA) along with polylactide may be used to prepare a biodegradable copolymer to be used as implantations. Fabrication of biopolymer/carbon nanotube composite has been reported using PGA fiber as one of the constituents of the composite. Modified porous PGA scaffolds has been fabricated.
medical device

Physical properties

Biodegradable polymer

Preparation Note

To achieve solubility in hexafluoroisopropanol, melt crystalline product and quench with liquid nitrogen to change material to amorphous form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preliminary report on the osteogenic potential of a biodegradable copolymer of polyactide (PLA) and polyglycolide (PGA)
Hollinger JO
Journal of Biomedical Materials Research Part A, 17(1) (1983)
Diopside modified porous polyglycolide scaffolds with improved properties
Feng P, et al.
Royal Society of Chemistry Advances, 5, 54822-54829 (2015)
Effect of dsDNA wrapped single-walled carbon nanotubes on the thermal and mechanical properties of polycaprolactone and polyglycolide fiber blend composites
Spearman SS
Polymer, 56(15), 476-481 (2015)
Fast Synthesis of High-Molecular-Weight Polyglycolide Using Diphenyl Bismuth Bromide as Catalyst
Lu Y, et al.
Macromolecular Chemistry and Physics, 216(4), 395-399 (2015)
Andrew K Capulli et al.
Biomaterials, 133, 229-241 (2017-04-27)
Tissue engineered scaffolds have emerged as a promising solution for heart valve replacement because of their potential for regeneration. However, traditional heart valve tissue engineering has relied on resource-intensive, cell-based manufacturing, which increases cost and hinders clinical translation. To overcome

Articles

Interest in utilizing biodegradable polymers for biomedical applications has grown since the 1960s.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service