Skip to Content
Merck
All Photos(1)

Key Documents

456373

Sigma-Aldrich

2,5-Dibromo-3-hexylthiophene

97%

Synonym(s):

2,5-Dibromo-3-hex-1-ylthiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14Br2S
CAS Number:
Molecular Weight:
326.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.557 (lit.)

density

1.521 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCc1cc(Br)sc1Br

InChI

1S/C10H14Br2S/c1-2-3-4-5-6-8-7-9(11)13-10(8)12/h7H,2-6H2,1H3

InChI key

NSYFIAVPXHGRSH-UHFFFAOYSA-N

General description

2,5-Dibromo-3-hexylthiophene is a 2,5 coupled conductive polymer with conjugated polythiophene based system, which has a controllable band gap.[1][2]

Application

Conducting polymer precursor.[3]
It can be used as a monomer with 5,5′-dibromo-3,3′-dihexyl-2,2′-bithiophene, which can synthesize regioregular-P3HT-regiosymmetric-P3HT (a diblock polymer) for organic electronics based applications.[4] It can also be used in the synthesis of P3HT, which can be potentially used for photocatalytic applications.[5]

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charge Transport in Conjugated Polymers with Pendent Stable Radical Groups.
Zhang Y, et al.
Chemistry of Materials (2018)
Paulina Powroznik et al.
Sensors (Basel, Switzerland), 20(2) (2020-01-19)
In the present work, we report the use of regioregular poly(3-hexyltiophene) polymer (RR-P3HT) as a potential light-activated material for sensing the chemical nerve agent simulant dimethyl methylphosphonate (DMMP). The electrical response of thick films of RR-P3HT, deposited by spray-coating method
Evidence of Ni (0) complex diffusion during grignard metathesis polymerization of 2, 5-dibromo-3-hexylthiophene.
Achord BC and Rawlins JW
Macromolecules, 42(22), 8634-8639 (2009)
Uniform electroactive fibre-like micelle nanowires for organic electronics.
Li X, et al.
Nature Communications, 8(22), 15909-15909 (2017)
Xiaoyu Li et al.
Nature communications, 8, 15909-15909 (2017-06-27)
Micelles formed by the self-assembly of block copolymers in selective solvents have attracted widespread attention and have uses in a wide variety of fields, whereas applications based on their electronic properties are virtually unexplored. Herein we describe studies of solution-processable

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service