454494
tert-Butyl tetraisopropylphosphorodiamidite
95%
Synonym(s):
1,1-Dimethylethyl N,N,N′,N′-tetrakis(1-methylethyl)phosphorodiamidite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[[(CH3)2CH]2N]2POC(CH3)3
CAS Number:
Molecular Weight:
304.45
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
form
solid
mp
67-74 °C (lit.)
functional group
amine
storage temp.
2-8°C
SMILES string
CC(C)N(C(C)C)P(OC(C)(C)C)N(C(C)C)C(C)C
InChI
1S/C16H37N2OP/c1-12(2)17(13(3)4)20(19-16(9,10)11)18(14(5)6)15(7)8/h12-15H,1-11H3
InChI key
PSFUFSJCFKUIIG-UHFFFAOYSA-N
General description
Tert-butyl tetraisopropylphosphorodiamidite is an effective ligand for palladium-catalyzed Buchwald–Hartwig amination of aryl- and heteroaryl chlorides and bromides widely used in the synthesis of industrially significant aryl- and heteroarylamines due to its accessibility and air-stability.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Stéphane Dumas et al.
Investigative radiology, 45(10), 600-612 (2010-09-03)
The donor atoms that bind to gadolinium in contrast agents influence inner-sphere water exchange and electronic relaxation, both of which determine observed relaxivity. The effect of these molecular parameters on relaxivity is greatest when the contrast agent is protein bound.
Palladium-Catalysed Amination of Aryl-and Heteroaryl Halides Using tert-Butyl Tetraisopropylphosphorodiamidite as an Easily Accessible and Air-Stable Ligand.
Roiban G-D, et al.
European Journal of Organic Chemistry, 10, 2070-2076 (2014)
Palladium-Catalysed Amination of Aryl- and Heteroaryl Halides Using tert-Butyl Tetraisopropylphosphorodiamidite as an Easily Accessible and Air-Stable Ligand
Roiban GD, et al.
European Journal of Organic Chemistry, 2014, 2070-2076 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service