Skip to Content
Merck
All Photos(2)

Documents

452904

Sigma-Aldrich

Sodium borohydride solution

~12 wt. % in 14 M NaOH

Synonym(s):

Sodium hydroborate, Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

Quality Level

reaction suitability

reagent type: reductant

concentration

~12 wt. % in 14 M NaOH

density

1.375 g/mL at 25 °C

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Alkaline sodium borohydride solution can be used as an efficient reagent for the generation of hydrogen via hydrolysis in the presence of various catalysts such as γ-Al2O3 supported cobalt catalysts, carbon supported cobalt catalysts, graphene oxide particles supported with cobalt nanoparticles and Ni45Au45Co10 trimetallic nanoparticles.

Legal Information

Product of Ascensus

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrogen generation utilizing alkaline sodium borohydride solution and supported cobalt catalyst
Ye W, et al.
Journal of Power Sources, 164(2), 544-548 (2007)
Carbon-supported cobalt catalyst for hydrogen generation from alkaline sodium borohydride solution
Xu D, et al.
Journal of Power Sources, 182(2), 616-620 (2008)
Efficient hydrolysis of alkaline sodium borohydride catalyzed by cobalt nanoparticles supported on three-dimensional graphene oxide
Wang J, et al.
Materials Research Bulletin, 95(2), 204-210 (2017)
Synthesis of Ni/Au/Co trimetallic nanoparticles and their catalytic activity for hydrogen generation from alkaline sodium borohydride aqueous solution
Jiao C, et al.
Royal Society of Chemistry Advances, 5(43), 34364-34371 (2015)
Meganne L Christian et al.
ACS nano, 6(9), 7739-7751 (2012-08-10)
Owing to its high storage capacity (10.8 mass %), sodium borohydride (NaBH(4)) is a promising hydrogen storage material. However, the temperature for hydrogen release is high (>500 °C), and reversibility of the release is unachievable under reasonable conditions. Herein, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service