Skip to Content
Merck
All Photos(2)

Key Documents

450855

Sigma-Aldrich

Gadolinium(III) chloride hydrate

99.99% trace metals basis

Synonym(s):

Gadolinium chloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GdCl3 · xH2O
CAS Number:
Molecular Weight:
263.61 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
solid
Pricing and availability is not currently available.

Assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: gadolinium

technique(s)

NMR: suitable

impurities

≤200.0 ppm Trace Rare Earth Analysis

density

2.424 g/mL at 25 °C (lit.)

SMILES string

O.Cl[Gd](Cl)Cl

InChI

1S/3ClH.Gd.H2O/h3*1H;;1H2/q;;;+3;/p-3

InChI key

SXVCBXHAHBCOMV-UHFFFAOYSA-K

Application

Employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yingying Li et al.
Frontiers in chemistry, 8, 832-832 (2020-11-12)
The rare-earth nanocrystals containing Er3+ emitters offer very promising tools for imaging applications, as they can not only exhibit up-conversion luminescence but also down-conversion luminescence in the second near-infrared window (NIR II). Doping non-lanthanide cations into host matrix was demonstrated
Silvio Aime et al.
Inorganic chemistry, 36(14), 2992-3000 (1997-07-02)
The synthesis of three triazamacrocycles containing the pyridine moiety and three acetate pendant arms (PCTA) is reported. The three systems differ due to the number of carbon atoms in the macrocyclic ring forming ligands PCTA-[12], -[13], and -[14], endowed with
MyeongJu Moon et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 17(4), 497-503 (2015-01-22)
In this study, we synthesized hyaluronic acid-conjugated gadolinium (HA-diethylene triamine pentaacetic acid (DTPA)-Gd) and evaluated as hepatocyte-specific magnetic resonance imaging (MRI) contrast agent for the diagnosis of hepatic metastasis. We conducted Fourier transform (FT)-IR analysis to determine the conjugation of

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service