Skip to Content
Merck
All Photos(1)

Documents

449679

Sigma-Aldrich

Barium isopropoxide

powder, 99% trace metals basis

Synonym(s):

Barium diisopropoxide, Barium isopropoxide, Diisopropoxybarium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(OCH(CH3)2)2
CAS Number:
Molecular Weight:
255.50
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99% trace metals basis

form

powder
solid

reaction suitability

core: barium

mp

200 °C (dec.) (lit.)

SMILES string

CC(C)O[Ba]OC(C)C

InChI

1S/2C3H7O.Ba/c2*1-3(2)4;/h2*3H,1-2H3;/q2*-1;+2

InChI key

CPUJSIVIXCTVEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For preparation of bimetallic Ba-Nb and Ba-Zr alkoxides. Asymmetric aldol reactions are promoted by a novel catalyst prepared from Ba(O-i-Pr)2.
It may be used as a catalyst for one pot preparation of symmetrical 1,5-diketones. Sol gel synthesis of ternary compound, Ba3 Li 2Ti8 O20 can be carried out by precursor reactants, barium isopropoxide, titanium isopropoxide and lithium acetate.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Self-heat. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Turevskaya, E.P. et al.
Polyhedron, 14, 1531-1531 (1995)
Yamada, Y.M.A. Shibasaki, M.
Tetrahedron Letters, 39, 5561-5561 (1998)
One-pot synthesis of 1,5-diketones catalyzed by barium isopropoxide
Yanagisawa A, et al.
Tetrahedron, 63.35, 8581-8585 (2007)
Preparation of YBaCuO powder by sol-gel techniques using molecular precursors.
El Khokh N, et al.
Journal of Materials Science Letters, 8.7, 762-764 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service