Skip to Content
Merck
All Photos(1)

Documents

442178

Sigma-Aldrich

p-Tolylmagnesium bromide solution

1.0 M in THF

Synonym(s):

4-Methylphenylmagnesium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4MgBr
CAS Number:
Molecular Weight:
195.34
Beilstein:
636491
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

1.0 M in THF

bp

65-67 °C

density

1.002 g/mL at 25 °C

SMILES string

Cc1ccc([Mg]Br)cc1

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

p-Tolylmagnesium bromide is a common Grignard reagent. It can also be used in a variety of cross-coupling reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-catalyzed cross coupling of Grignard reagents with in situ-derived enol phosphates.
Miller JA
Tetrahedron Letters, 43(39), 7111-7114 (2002)
Dinuclear Iron Complex-Catalyzed Cross-Coupling of Primary Alkyl Fluorides with Aryl Grignard Reagents.
Mo Z, et al.
Organometallics, 31(18), 6518-6521 (2012)
Palladium-and Nickel-Catalyzed Kumada Cross-Coupling Reactions of gem-Difluoroalkenes and Monofluoroalkenes with Grignard Reagents.
Dai W
The Journal of Organic Chemistry, 79(21), 10537-10546 (2014)
Takuji Hatakeyama et al.
Journal of the American Chemical Society, 131(33), 11949-11963 (2009-07-31)
Combinations of N-heterocyclic carbenes (NHCs) and fluoride salts of the iron-group metals (Fe, Co, and Ni) have been shown to be excellent catalysts for the cross-coupling reactions of aryl Grignard reagents (Ar(1)MgBr) with aryl and heteroaryl halides (Ar(2)X) to give

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service