Skip to Content
Merck
All Photos(1)

Documents

441287

Sigma-Aldrich

(1S)-(+)-2-Azabicyclo[2.2.1]hept-5-en-3-one

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

optical activity

[α]20/D +565°, c = 1 in chloroform

optical purity

ee: 99% (HPLC)

mp

94-97 °C (lit.)

SMILES string

O=C1N[C@H]2C[C@@H]1C=C2

InChI

1S/C6H7NO/c8-6-4-1-2-5(3-4)7-6/h1-2,4-5H,3H2,(H,7,8)/t4-,5+/m0/s1

InChI key

DDUFYKNOXPZZIW-CRCLSJGQSA-N

Application

This lactam and its antipode are precursors to the enantiomers of the carbocyclic nucleoside carbovir. It has been demonstrated that (-)-carbovir is a potent inhibitor of HIV-1 and based on clinical trials it could become an alternative to AZT. AZT is a trademark of The Wellcome Foundation Ltd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E L White et al.
Biochemical and biophysical research communications, 161(2), 393-398 (1989-06-15)
Carbocylic 2',3'-didehydro-2',3'-dideoxyguanosine (Carbovir; NSC 614846) is an antiretroviral agent which may be useful in the treatment of AIDS. We have synthesized the 5'-triphosphate of Carbovir and examined its ability to inhibit human immunodeficiency virus type 1 (HIV-1) reverse transcriptase (EC
Jung. M.E. Rhee, H.
The Journal of Organic Chemistry, 59, 4719-4719 (1994)
Campbell, J.A. et al.
The Journal of Organic Chemistry, 60, 4602-4602 (1995)
Evans, C.T. et al.
Journal of the Chemical Society. Perkin Transactions 1, 589-589 (1992)
R Vince et al.
Journal of medicinal chemistry, 33(1), 17-21 (1990-01-01)
(+-)-cis-[4-[(2,5-Diamino-6-chloropyrimidinyl)amino]-2- cyclopentenyl]carbinol (5a) was synthesized from 2-amino-4,6-dichloropyrimidine and cis-4-(hydroxymethyl)cyclopentenylamine (2a) by subsequent preparation of the 5-[(4-chlorophenyl)azo] derivative of the resulting pyrimidine (3a) and reduction of the azo moiety with zinc and acetic acid. The carbocyclic analogue of 2',3'-didehydro-2',3'-dideoxy 2-amino-6-chloropurine (6a)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service