Skip to Content
Merck
All Photos(1)

Key Documents

439614

Sigma-Aldrich

Ytterbium(III) chloride

anhydrous, powder, 99.9%

Synonym(s):

Ytterbium chloride, Ytterbium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YbCl3
CAS Number:
Molecular Weight:
279.40
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

99.9%

form

powder

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

875 °C (lit.)

SMILES string

Cl[Yb](Cl)Cl

InChI

1S/3ClH.Yb/h3*1H;/q;;;+3/p-3

InChI key

CKLHRQNQYIJFFX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Applied with NiCl2 to form a very effective catalyst for the reductive dehalogenation of aryl halides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhang, Y. et al.
Synthetic Communications, 27, 4327-4327 (1997)
W Shepard et al.
Acta crystallographica. Section D, Biological crystallography, 56(Pt 10), 1288-1303 (2000-09-22)
The basic theory and principles of the multiple-wavelength anomalous solvent-contrast (MASC) method are introduced as a contrast-variation technique for generating low-resolution crystallographic phase information on the envelope of a macromolecule. Experimental techniques and practical considerations concerning the choice of anomalous
Heng Qin et al.
Physical chemistry chemical physics : PCCP, 19(29), 19159-19167 (2017-07-14)
Efficient enhancement of photoluminescence in rare-earth activated upconversion materials is of great significance for their practical applications in various fields. In this work, three-dimensional mesoporous gold films were fabricated by a low-cost and facile dealloying approach to improve the upconversion
D M Albin et al.
Journal of animal science, 79(5), 1250-1258 (2001-05-26)
The apparent ileal digestibilities of amino acids and rate of passage were evaluated in pigs (BW = 78.3 +/- 7.4 kg) fed a semipurified diet. The pigs were fed 1.82, 2.73, or 3.65 kg DMI/d. The highest level of feed
Peng Zhang et al.
Environmental science & technology, 46(3), 1834-1841 (2011-12-24)
With the increasing utilization of nanomaterials, there is a growing concern for the potential environmental and health effects of them. To assess the environmental risks of nanomaterials, better knowledge about their fate and toxicity in plants are required. In this

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service